Learn More
Strem, An Ascensus Company CAS# 12065-11-1. 1g. Germanium(IV) selenide (99.999%-Ge) PURATREM. MFCD00016116
SDP
Catalog No. 5090115064
Encompass
CAS# 12065-11-1. 1g. Germanium(IV) selenide (99.999%-Ge) PURATREM. MFCD00016116. Molecular Weight: 230.51. Molecular Formula: GeSe2. Color/form: orange xtl. Strem# 93-3224. http://www.strem.com/catalog/v/93-3224/
Catalog No. 50-901-15064 Supplier Strem, An Ascensus Company Supplier No. 9332241G
CAS# 12065-11-1. 1g. Germanium(IV) selenide (99.999%-Ge) PURATREM. MFCD00016116. Molecular Weight: 230.51. Molecular Formula: GeSe2. Color/form: orange xtl. Strem# 93-3224. http://www.strem.com/catalog/v/93-3224/