Learn More
Grainger Germicidal Bleach, 121 oz Cleaner Container Size, Jug Cleaner Container Type, Unscented Fragrance
Catalog No. NC0566218
Encompass_Preferred
Germicidal bleach is more concentrated than standard bleach, so less product is needed to create a solution for disinfecting surfaces. Apply it to hard nonporous surfaces to eliminate mold, odors, and a broad range of pathogens, including bacteria, fungi, and viruses. The surfaces should be cleaned before applying bleach. Germicidal bleach is commonly used in healthcare facilities, offices, schools, kitchens, and other commercial sites.
This item is not returnable. View return policy
Catalog No. NC0566218 Supplier Grainger Supplier No. 41H893
Germicidal bleach is more concentrated than standard bleach, so less product is needed to create a solution for disinfecting surfaces. Apply it to hard nonporous surfaces to eliminate mold, odors, and a broad range of pathogens, including bacteria, fungi, and viruses. The surfaces should be cleaned before applying bleach. Germicidal bleach is commonly used in healthcare facilities, offices, schools, kitchens, and other commercial sites.