Learn More
Bulbman Inc USHIO G36T5L GERMICIDAL 39W 33IN
SDP
Catalog No. NC9136730
Encompass_Preferred
Germicidal Lamp, 39w, instant start lamp, The UV light bulb is tubular with a glass diameter of 0.675 in., an FA8 base and an overall length of 33.2 in., It is a low ozone light bulb rated at 9000 life hours
Catalog No. NC9136730 Supplier Bulbman Inc Supplier No. 119003
Germicidal Lamp, 39w, instant start lamp, The UV light bulb is tubular with a glass diameter of 0.675 in., an FA8 base and an overall length of 33.2 in., It is a low ozone light bulb rated at 9000 life hours