Learn More
MSC Detco Disinfectant Cleaner, Concentrated, Detergent Disinfectant, Lemon Scent, 15 Gal
Catalog No. 502816385
Encompass
Germ-A-Cide 64 is a concentrated disinfectant and cleaner designed for use on washable surfaces. Provided in a 15-gallon drum, it offers effective cleaning and disinfection in a single solution.
  • Concentrated detergent disinfectant formula
  • Suitable for a wide range of washable surfaces
  • Lemon scented for a fresh aroma
  • Requires a dilution of 2 oz. per gallon
  • Non-acidic composition
  • Low VOC content of 0.1%
  • Biodegradable for environmental consideration
Catalog No. 50-281-6385 Supplier MSC Supplier No. 33702523
Germ-A-Cide 64 is a concentrated disinfectant and cleaner designed for use on washable surfaces. Provided in a 15-gallon drum, it offers effective cleaning and disinfection in a single solution.
  • Concentrated detergent disinfectant formula
  • Suitable for a wide range of washable surfaces
  • Lemon scented for a fresh aroma
  • Requires a dilution of 2 oz. per gallon
  • Non-acidic composition
  • Low VOC content of 0.1%
  • Biodegradable for environmental consideration