Learn More
MSC Detco Germ-A-Cide 64, Disinfectant/Cleaner, 55 gal, Lemon, 1 ea
Catalog No. 502816386
Encompass
Detco Germ-A-Cide 64 is a concentrated liquid disinfectant and cleaner designed for effective sanitation across various washable surfaces in institutional and commercial settings.
  • Disinfectant liquid form
  • Packaged in a 55 gal drum
  • Lemon scented for a fresh aroma
  • Recommended dilution of 2 oz/gal
  • Effective on washable surfaces
  • Features Alkyl Dimethyl Benzyl Ammonium Chloride as primary chemical
  • VOC content of 0.1%
  • Biodegradable formula
  • Non-acidic composition
  • Made in the United States
Catalog No. 50-281-6386 Supplier MSC Supplier No. 33702531
Detco Germ-A-Cide 64 is a concentrated liquid disinfectant and cleaner designed for effective sanitation across various washable surfaces in institutional and commercial settings.
  • Disinfectant liquid form
  • Packaged in a 55 gal drum
  • Lemon scented for a fresh aroma
  • Recommended dilution of 2 oz/gal
  • Effective on washable surfaces
  • Features Alkyl Dimethyl Benzyl Ammonium Chloride as primary chemical
  • VOC content of 0.1%
  • Biodegradable formula
  • Non-acidic composition
  • Made in the United States