Learn More
MSC PortionPac Germicidal Cleaner, 7 oz Packet, Disinfectant, Ultra-Concentrated, 24/Pk.
Catalog No. 502868964
Encompass
This germicidal cleaner is an ultra-concentrated, EPA Registered solution supplied in 7 oz packets, designed to function as an effective disinfectant for a wide range of surfaces.
  • Efficacy as a bactericide, fungicide, virucide, and mildewcide
  • Effective against Staphylococcus aureus, Salmonella enterica, Pseudomonas aeruginosa, MRSA, Influenza Virus Type A2, Avian Influenza Virus, HIV Type 1, Trichophyton interdigitale, Aspergillus niger, and Candida albicans
  • Suitable for use on floors, walls, toilets, tables, mirrors, and appliance exteriors
  • Each packet makes 7 gal. of cleaning solution
  • Ultra-concentrated formula
  • Unscented liquid
Catalog No. 50-286-8964 Supplier MSC Supplier No. 38688966
This germicidal cleaner is an ultra-concentrated, EPA Registered solution supplied in 7 oz packets, designed to function as an effective disinfectant for a wide range of surfaces.
  • Efficacy as a bactericide, fungicide, virucide, and mildewcide
  • Effective against Staphylococcus aureus, Salmonella enterica, Pseudomonas aeruginosa, MRSA, Influenza Virus Type A2, Avian Influenza Virus, HIV Type 1, Trichophyton interdigitale, Aspergillus niger, and Candida albicans
  • Suitable for use on floors, walls, toilets, tables, mirrors, and appliance exteriors
  • Each packet makes 7 gal. of cleaning solution
  • Ultra-concentrated formula
  • Unscented liquid