Learn More
GEXPRO GERMICIDAL LAMP G30T8 36IN
Catalog No. NC2207372
Encompass_Preferred
  • Engineered for effective inactivation of microorganisms.
  • Operating wattage of 30 W.
  • Emits short-wave UV radiation at 253.7 nm.
  • Durable design ensures consistent UV output.
  • Average rated life of approximately 8,000 hours.
  • Standard T8 tubular shape with a Medium Bi-Pin (G13) base.
  • Compatible with a variety of germicidal systems.
  • Primary applications include air disinfection and water purification.
  • Carefully designed to disrupt the DNA of harmful microorganisms.
  • Safety warning: Emits harmful UV-C radiation; avoid exposure to eyes and skin.
This item is not returnable. View return policy
Catalog No. NC2207372 Supplier GEXPRO Supplier No. G30T8
  • Engineered for effective inactivation of microorganisms.
  • Operating wattage of 30 W.
  • Emits short-wave UV radiation at 253.7 nm.
  • Durable design ensures consistent UV output.
  • Average rated life of approximately 8,000 hours.
  • Standard T8 tubular shape with a Medium Bi-Pin (G13) base.
  • Compatible with a variety of germicidal systems.
  • Primary applications include air disinfection and water purification.
  • Carefully designed to disrupt the DNA of harmful microorganisms.
  • Safety warning: Emits harmful UV-C radiation; avoid exposure to eyes and skin.