Learn More
Roboz Surgical GERMINATOR 500 BEAD STERALIZER
SDP
Catalog No. NC9780119
Encompass
The GERMINATOR 500 is a simple, inexpensive way to quickly decontaminate your micro dissecting instruments between procedures. The unique stainless steel glass bead bath remains at a constant 500 degrees F, allowing you to insert the tips of your instruments whenever you wish to decontaminate them. And it’s fast...most micro-dissecting instruments decontaminate within 15 seconds! No messy chemicals and no need for open flames in the lab. Easy to use ◾Fast, effective decontamination ◾Small footprint saves bench space ◾Proven durable design
Catalog No. NC9780119 Supplier Roboz Surgical Supplier No. DS401
The GERMINATOR 500 is a simple, inexpensive way to quickly decontaminate your micro dissecting instruments between procedures. The unique stainless steel glass bead bath remains at a constant 500 degrees F, allowing you to insert the tips of your instruments whenever you wish to decontaminate them. And it’s fast...most micro-dissecting instruments decontaminate within 15 seconds! No messy chemicals and no need for open flames in the lab. Easy to use ◾Fast, effective decontamination ◾Small footprint saves bench space ◾Proven durable design