Learn More
Neuvitro CE certified German glass coverslips for cell culture and confocal /IHC imaging, 25mm diameter #1.5 thick 100 /pk untreated
Catalog No. NC0530564
Encompass_Preferred
CE certified cell culture tested German glass coverslips for cell culture and confocal /IHC imaging, 25mm diameter #1.5 thickness 100 pieces/pk untreated. Pretreatment is required prior to cell culture applications. Shelf life 5 years, store at room temperature in dry.
Catalog No. NC0530564 Supplier Neuvitro Supplier No. GG251.5
CE certified cell culture tested German glass coverslips for cell culture and confocal /IHC imaging, 25mm diameter #1.5 thickness 100 pieces/pk untreated. Pretreatment is required prior to cell culture applications. Shelf life 5 years, store at room temperature in dry.