Learn More
Cayman Chemical GernylgernoIc AcId 100UG
SDP
Catalog No. 502895661
Encompass_Preferred
An isoprenoid with anticancer activity; induces apoptosis in Huh7 and PLC/PRF/5 human hepatoma cells and MLE-10 transformed mouse hepatocytes from 1-20 µM; induces apoptosis in Huh7 cells via loss of the mitochondrial membrane potential and activation of ICE and CPP32 at 10 µM; inhibits LSD1 (IC50 = 46.97 µM)
Catalog No. 50-289-5661 Supplier Cayman Chemical Supplier No. 9003474100 UG
An isoprenoid with anticancer activity; induces apoptosis in Huh7 and PLC/PRF/5 human hepatoma cells and MLE-10 transformed mouse hepatocytes from 1-20 µM; induces apoptosis in Huh7 cells via loss of the mitochondrial membrane potential and activation of ICE and CPP32 at 10 µM; inhibits LSD1 (IC50 = 46.97 µM)