Learn More
Cayman Chemical Geranylgeranylacetone, Purity: ≥98% (mixture of isomers), CAS: 6809-52-5, 50 mg
SDP
Catalog No. 502982483
Encompass_Preferred
An inducer of Hsp expression that increases Hsp70, Hsp22, Hsp27, Hsp90, and Hsp105 levels in various cells and tissues; orally bioavailable and has diverse effects in vivo, including hepatoprotective, neuroprotective, and antiulcerative effects
Catalog No. 50-298-2483 Supplier Cayman Chemical Supplier No. 2021850 MG
An inducer of Hsp expression that increases Hsp70, Hsp22, Hsp27, Hsp90, and Hsp105 levels in various cells and tissues; orally bioavailable and has diverse effects in vivo, including hepatoprotective, neuroprotective, and antiulcerative effects