Learn More
Gerstel GERSTEL 10 µL Syringe, Fixed Needle, Stainless Steel Plunger, 10 µL
SDP
Catalog No. NC1085276
Encompass
The GERSTEL 10 µL Syringe is engineered for precision liquid handling in automated scientific environments, making it ideal for chromatographic applications. It features a fixed needle made of stainless steel and a specially designed conical tip for effective septum penetration, ensuring sample integrity during repetitive injections. This syringe's optimized design for use with GERSTEL MultiPurpose Sampler (MPS) robotic systems enhances reliability and reproducibility, setting it apart from standard syringes.
Catalog No. NC1085276 Supplier Gerstel Supplier No. 10011101400
The GERSTEL 10 µL Syringe is engineered for precision liquid handling in automated scientific environments, making it ideal for chromatographic applications. It features a fixed needle made of stainless steel and a specially designed conical tip for effective septum penetration, ensuring sample integrity during repetitive injections. This syringe's optimized design for use with GERSTEL MultiPurpose Sampler (MPS) robotic systems enhances reliability and reproducibility, setting it apart from standard syringes.