Learn More
Med Vet International Equispon Absorbable Get Foam, 80mm x 50mm x 1mm, J1275A, Each
Catalog No. 5000495717
Encompass_Preferred
A local hemostatic sponge for use in surgical procedures that involve venous bleeding and oozing where standard hemostats are difficult or impractical• Achieves hemostatis rapidly in a few minutes. • Can be trimmed to size or sutured in place• Absorbs 50 times its own weight• Uniform porosity• Can use with medications and not affect its hemostatic properties• Sterile double packaged80mm x 50mm x 1mm strip
Catalog No. 50-004-95717 Supplier Med Vet International Supplier No. JOR1275A
A local hemostatic sponge for use in surgical procedures that involve venous bleeding and oozing where standard hemostats are difficult or impractical• Achieves hemostatis rapidly in a few minutes. • Can be trimmed to size or sutured in place• Absorbs 50 times its own weight• Uniform porosity• Can use with medications and not affect its hemostatic properties• Sterile double packaged80mm x 50mm x 1mm strip