Learn More
Aviva Systems Biology Corp GET4 Antibody - N-terminal region (ARP61198_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61198_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GET4. It was validated on Western Blot. . Alternate names: CEE, CGI-20, TRC35, C7orf20ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-287-291