Learn More
CELLTREAT Scientific Products Glass Fiber Filter Disk, Prefilter, Binder Free, 1.0m, 50mm, Sterile, 50/CS
SDP
Supplier:  CELLTREAT Scientific Products 230730
Encompass_Preferred
CELLTREAT Scientific Products Glass Fiber Filter Disk, Prefilter, Binder Free, 1.0¿m, 50mm Diameter. Glass Fiber Filter Disks are an ideal prefilter to prevent clogging when filtering viscous or particulate-laden solutions with CELLTREAT Scientific Products vacuum Filter Systems and Bottle Top Filters. Increase filtration speed by removing larger particles prior to sterilizing filtration. Glass Fiber material. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, individually wrapped, 50/case.
Catalog No. 50-153-5139