Learn More
CELLTREAT Scientific Products Glass Fiber Filter Disk, Prefilter, Binder Free, 1.0m, 70mm, Sterile, 50/CS
SDP
Catalog No. 501535140
Encompass_Preferred
CELLTREAT Scientific Products Glass Fiber Filter Disk, Prefilter, Binder Free, 1.0¿m, 70mm Diameter. Glass Fiber Filter Disks are an ideal prefilter to prevent clogging when filtering viscous or particulate-laden solutions with CELLTREAT Scientific Products vacuum Filter Systems and Bottle Top Filters. Increase filtration speed by removing larger particles prior to sterilizing filtration. Glass Fiber material. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, individually wrapped, 50/case.
Catalog No. 50-153-5140 Supplier CELLTREAT Scientific Products Supplier No. 230732
CELLTREAT Scientific Products Glass Fiber Filter Disk, Prefilter, Binder Free, 1.0¿m, 70mm Diameter. Glass Fiber Filter Disks are an ideal prefilter to prevent clogging when filtering viscous or particulate-laden solutions with CELLTREAT Scientific Products vacuum Filter Systems and Bottle Top Filters. Increase filtration speed by removing larger particles prior to sterilizing filtration. Glass Fiber material. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, individually wrapped, 50/case.