Learn More
MSC GF Gage Pipe Thread Plug Gage, Tapered, 1/4-18, Class L-1, Single End
Catalog No. 5000528768
Encompass
This Pipe Thread Plug Gage from GF Gage is designed for precision measurement of internal tapered threads. It features a 1/4-18 thread size, Class L-1 accuracy, and a single-end design for efficient use.
  • Tapered design for accurate thread inspection.
  • 1/4-18 thread size for standard applications.
  • Class L-1 for precise measurement.
  • Single end configuration includes a size 3 handle.
  • Ideal for NPTF-L1 thread types.
  • Made in the United States.
Catalog No. 50-005-28768 Supplier MSC Supplier No. 55337950
This Pipe Thread Plug Gage from GF Gage is designed for precision measurement of internal tapered threads. It features a 1/4-18 thread size, Class L-1 accuracy, and a single-end design for efficient use.
  • Tapered design for accurate thread inspection.
  • 1/4-18 thread size for standard applications.
  • Class L-1 for precise measurement.
  • Single end configuration includes a size 3 handle.
  • Ideal for NPTF-L1 thread types.
  • Made in the United States.