Learn More
MSC GF Gage Pipe Thread Plug Gage, Tapered, 2-11-1/2, Class L-1, Single End
Catalog No. 5000527649
Encompass
This pipe thread plug gage is designed for precise measurement of tapered internal threads. It features a 2-11-1/2 thread size and Class L-1 accuracy, ensuring reliable inspection results. The single-ended design includes a handle for ease of use.
  • Tapered design for accurate thread measurement
  • 2-11-1/2 thread size for specific applications
  • Class L-1 for precise gaging
  • Single end configuration for focused application
  • Includes handle for easy operation
Catalog No. 50-005-27649 Supplier MSC Supplier No. 55336770
This pipe thread plug gage is designed for precise measurement of tapered internal threads. It features a 2-11-1/2 thread size and Class L-1 accuracy, ensuring reliable inspection results. The single-ended design includes a handle for ease of use.
  • Tapered design for accurate thread measurement
  • 2-11-1/2 thread size for specific applications
  • Class L-1 for precise gaging
  • Single end configuration for focused application
  • Includes handle for easy operation