Learn More
MSC GF Gage Pipe Thread Plug Gage, Tapered, 3/8-18, Class L-1
Catalog No. 5000535821
Encompass
This precision pipe thread plug gage is designed for accurately measuring 3/8-18 tapered pipe threads. It features a Class L-1 rating and a single-end configuration for reliable and consistent gaging in industrial applications.
  • Tapered design for pipe threads.
  • Measures 3/8-18 thread size.
  • Class L-1 rating ensures precision.
  • Single end configuration.
Catalog No. 50-005-35821 Supplier MSC Supplier No. 55337976
This precision pipe thread plug gage is designed for accurately measuring 3/8-18 tapered pipe threads. It features a Class L-1 rating and a single-end configuration for reliable and consistent gaging in industrial applications.
  • Tapered design for pipe threads.
  • Measures 3/8-18 thread size.
  • Class L-1 rating ensures precision.
  • Single end configuration.