Learn More
MSC GF Gage Plug Thread Gage, 1/2-28 Thread, 3B Class, Go/No Go, Double End
Catalog No. 5000529723
Encompass
This plug thread gage is designed for precise measurement of 1/2-28 threads with a 3B class. It features a double-ended design with both Go and No Go functionalities, ensuring accurate and reliable thread inspection. Made from hardened tool steel, this gage is durable and includes a handle for ease of use.
  • Go pitch diameter of 0.4768 inches
  • No go pitch diameter of 0.4804 inches
  • Handle is included
  • Manufactured from hardened tool steel
  • Double-ended design for comprehensive thread checking
  • Ensures accurate Go and No Go measurements
  • Made in the United States
Catalog No. 50-005-29723 Supplier MSC Supplier No. 05394291
This plug thread gage is designed for precise measurement of 1/2-28 threads with a 3B class. It features a double-ended design with both Go and No Go functionalities, ensuring accurate and reliable thread inspection. Made from hardened tool steel, this gage is durable and includes a handle for ease of use.
  • Go pitch diameter of 0.4768 inches
  • No go pitch diameter of 0.4804 inches
  • Handle is included
  • Manufactured from hardened tool steel
  • Double-ended design for comprehensive thread checking
  • Ensures accurate Go and No Go measurements
  • Made in the United States