Learn More
MSC GF Gage Plug Thread Gage, #10-24 Thread, Go, High-Speed Steel, Titanium Nitride Coated
Catalog No. 5000528179
Encompass
This plug thread gage is designed for checking #10-24 threads. It is a "Go" type gage, ensuring that threads meet minimum material limits. Constructed from high-speed steel and finished with a titanium nitride coating, it offers durability and extended tool life. This single-end gage does not include a handle.
  • Designed for #10-24 threads
  • Ensures threads meet minimum material limits with a "Go" type gage
  • Constructed from high-speed steel for durability
  • Titanium nitride coating provides extended tool life
  • Single-end design
  • Does not include a handle
  • Go pitch diameter of 0.1629 inch
  • Supports 2B and 3B classification
Catalog No. 50-005-28179 Supplier MSC Supplier No. 55312771
This plug thread gage is designed for checking #10-24 threads. It is a "Go" type gage, ensuring that threads meet minimum material limits. Constructed from high-speed steel and finished with a titanium nitride coating, it offers durability and extended tool life. This single-end gage does not include a handle.
  • Designed for #10-24 threads
  • Ensures threads meet minimum material limits with a "Go" type gage
  • Constructed from high-speed steel for durability
  • Titanium nitride coating provides extended tool life
  • Single-end design
  • Does not include a handle
  • Go pitch diameter of 0.1629 inch
  • Supports 2B and 3B classification