Learn More
MSC GF Gage Plug Thread Gage, #10-32 Thread, 2B & 3B Class, Single End, Go
Catalog No. 5000512907
Encompass
This plug thread gage is designed for #10-32 threads. It features 2B and 3B classes, is a single-end 'Go' gage, and has a Go Pitch Diameter of 0.1697 inches. Constructed from hardened tool steel, this gage is manufactured in the United States.
  • "Go" type gage for precise measurements.
  • Designed for #10-32 threads.
  • Constructed from hardened tool steel for durability.
  • Features 2B and 3B classifications.
  • Single-end design.
  • Go pitch diameter of 0.1697 "
  • Thread length of 13/32 "
  • Manufactured in the United States.
Catalog No. 50-005-12907 Supplier MSC Supplier No. 55306419
This plug thread gage is designed for #10-32 threads. It features 2B and 3B classes, is a single-end 'Go' gage, and has a Go Pitch Diameter of 0.1697 inches. Constructed from hardened tool steel, this gage is manufactured in the United States.
  • "Go" type gage for precise measurements.
  • Designed for #10-32 threads.
  • Constructed from hardened tool steel for durability.
  • Features 2B and 3B classifications.
  • Single-end design.
  • Go pitch diameter of 0.1697 "
  • Thread length of 13/32 "
  • Manufactured in the United States.