Learn More
MSC GF Gage Plug Thread Gage, #2-64 Thread, 2B Class, No Go
Catalog No. 5000534964
Encompass
This plug thread gage is designed for industrial applications to verify the thread size and class of #2-64 threads. It functions as a 'No Go' gage, ensuring the upper limit of the thread tolerance is not exceeded, which is critical for quality control in manufacturing and assembly processes.
  • Tests the no go condition of threads
  • Designed for #2-64 thread sizes
  • Constructed from hardened tool steel for durability
  • Features 2B thread classification
  • Single end configuration
  • No go pitch diameter of 0.07860 inch
  • Handle not included
Catalog No. 50-005-34964 Supplier MSC Supplier No. 55307334
This plug thread gage is designed for industrial applications to verify the thread size and class of #2-64 threads. It functions as a 'No Go' gage, ensuring the upper limit of the thread tolerance is not exceeded, which is critical for quality control in manufacturing and assembly processes.
  • Tests the no go condition of threads
  • Designed for #2-64 thread sizes
  • Constructed from hardened tool steel for durability
  • Features 2B thread classification
  • Single end configuration
  • No go pitch diameter of 0.07860 inch
  • Handle not included