Learn More
MSC GF Gage Plug Thread Gage, 3/4-10 Thread, 2B Class, Double End
Catalog No. 5000512906
Encompass
This plug thread gage is designed for inspecting 3/4-10 threads with a 2B classification. It features a double-end design, incorporating both Go and No Go gaging elements to ensure thread accuracy and compliance.
  • Made from hardened tool steel for durability.
  • Includes a handle for ease of use.
  • Features a Go pitch diameter of 0.6850 in and a No Go pitch diameter of 0.69270 in.
  • Comes with a certificate of compliance, ensuring proper tolerances and physical condition.
  • Manufactured in the United States.
Catalog No. 50-005-12906 Supplier MSC Supplier No. 55310379
This plug thread gage is designed for inspecting 3/4-10 threads with a 2B classification. It features a double-end design, incorporating both Go and No Go gaging elements to ensure thread accuracy and compliance.
  • Made from hardened tool steel for durability.
  • Includes a handle for ease of use.
  • Features a Go pitch diameter of 0.6850 in and a No Go pitch diameter of 0.69270 in.
  • Comes with a certificate of compliance, ensuring proper tolerances and physical condition.
  • Manufactured in the United States.