Learn More
MSC GF Gage Plug Thread Gage, 3/4-16, 2B, Double-End Go/No Go
Catalog No. 5000530300
Encompass
This plug thread gage is designed for precise measurement of 3/4-16 threads with a 2B class. It features a double-end design with both 'Go' and 'No Go' sides, made from durable hardened tool steel. A handle is included for ease of use, making it suitable for quality control and inspection tasks.
  • Double-end design with 'Go' and 'No Go' functionality.
  • Measures 3/4-16 threads.
  • Features 2B classification.
  • Constructed from hardened tool steel for durability.
  • Includes a handle for convenient operation.
  • Comes with a Certificate of Compliance.
  • Go pitch diameter is 0.7094 "
  • No Go pitch diameter is 0.71590 "
Catalog No. 50-005-30300 Supplier MSC Supplier No. 55310395
This plug thread gage is designed for precise measurement of 3/4-16 threads with a 2B class. It features a double-end design with both 'Go' and 'No Go' sides, made from durable hardened tool steel. A handle is included for ease of use, making it suitable for quality control and inspection tasks.
  • Double-end design with 'Go' and 'No Go' functionality.
  • Measures 3/4-16 threads.
  • Features 2B classification.
  • Constructed from hardened tool steel for durability.
  • Includes a handle for convenient operation.
  • Comes with a Certificate of Compliance.
  • Go pitch diameter is 0.7094 "
  • No Go pitch diameter is 0.71590 "