Learn More
MSC GF Gage Plug Thread Gage, 3/8-24 Thread, 3B Class, Double End
Catalog No. 5000528617
Encompass
This plug thread gage is designed for precise measurement of 3/8-24 threads with a 3B class fit. It features a double-ended configuration with both Go and No Go functions, ensuring accurate assessment of thread compliance.
  • Go and No Go functionality
  • 3/8-24 thread size
  • Hardened tool steel material
  • 3B class thread classification
  • Go pitch diameter: 0.3479 inch
  • No Go pitch diameter: 0.35160 inch
  • Double-ended design
  • Includes handle
Catalog No. 50-005-28617 Supplier MSC Supplier No. 55311252
This plug thread gage is designed for precise measurement of 3/8-24 threads with a 3B class fit. It features a double-ended configuration with both Go and No Go functions, ensuring accurate assessment of thread compliance.
  • Go and No Go functionality
  • 3/8-24 thread size
  • Hardened tool steel material
  • 3B class thread classification
  • Go pitch diameter: 0.3479 inch
  • No Go pitch diameter: 0.35160 inch
  • Double-ended design
  • Includes handle