Learn More
MSC GF Gage Plug Thread Gage, 3/8-24 Thread, STI 2B, No Go, Steel
Catalog No. 5000528616
Encompass
This is a GF Gage Plug Thread Gage designed for inspecting 3/8-24 STI 2B threads. It is a No Go gage, crafted from steel, and features a single-end design without a handle.
  • No go gage
  • 3/8-24 thread size
  • 2B classification
  • Screw thread insert (STI) type
  • Handle not included
  • Made from steel
  • Single end
Catalog No. 50-005-28616 Supplier MSC Supplier No. 55315337
This is a GF Gage Plug Thread Gage designed for inspecting 3/8-24 STI 2B threads. It is a No Go gage, crafted from steel, and features a single-end design without a handle.
  • No go gage
  • 3/8-24 thread size
  • 2B classification
  • Screw thread insert (STI) type
  • Handle not included
  • Made from steel
  • Single end