Learn More
MSC GF Gage Plug Thread Gage, 3/8-32 Thread, 2B Class, Go/No Go
Catalog No. 5000528626
Encompass
This Plug Thread Gage is designed for precise thread inspection, featuring a 3/8-32 thread with a 2B class. Its double-end design allows for both 'Go' and 'No Go' measurements, ensuring accuracy and reliability. Constructed from hardened tool steel, this gage is built for durability and long-lasting performance.
  • Designed for 3/8-32 thread measurements.
  • Features a 2B class for precision.
  • Double-end design for both go and no go measurements.
  • Constructed from hardened tool steel for durability.
  • Includes a handle for ease of use.
  • Go pitch diameter of 0.3547 "
  • No go pitch diameter of 0.3591 "
  • Manufactured in the United States.
Catalog No. 50-005-28626 Supplier MSC Supplier No. 55310114
This Plug Thread Gage is designed for precise thread inspection, featuring a 3/8-32 thread with a 2B class. Its double-end design allows for both 'Go' and 'No Go' measurements, ensuring accuracy and reliability. Constructed from hardened tool steel, this gage is built for durability and long-lasting performance.
  • Designed for 3/8-32 thread measurements.
  • Features a 2B class for precision.
  • Double-end design for both go and no go measurements.
  • Constructed from hardened tool steel for durability.
  • Includes a handle for ease of use.
  • Go pitch diameter of 0.3547 "
  • No go pitch diameter of 0.3591 "
  • Manufactured in the United States.