Learn More
MSC GF Gage Plug Thread Gage, #4-40 Thread, 2B Class, No Go, 0.0991 in Pitch Diameter, Single End
Catalog No. 5000527768
Encompass
The GF Gage Plug Thread Gage is designed for precise measurement of internal threads. This single-end, #4-40 thread gage with a 2B class and a 0.0991 in no go pitch diameter ensures accurate quality control and adherence to specifications.
  • Measures internal threads with a #4-40 thread size.
  • Features a 2B class for precise gaging.
  • "No Go" type for checking thread limits.
  • Constructed from high speed steel for durability.
  • Ensures accuracy with a 0.0991 in no go pitch diameter.
  • Single end configuration for specific applications.
  • TiN coating/finish for enhanced wear resistance.
Catalog No. 50-005-27768 Supplier MSC Supplier No. 55313050
The GF Gage Plug Thread Gage is designed for precise measurement of internal threads. This single-end, #4-40 thread gage with a 2B class and a 0.0991 in no go pitch diameter ensures accurate quality control and adherence to specifications.
  • Measures internal threads with a #4-40 thread size.
  • Features a 2B class for precise gaging.
  • "No Go" type for checking thread limits.
  • Constructed from high speed steel for durability.
  • Ensures accuracy with a 0.0991 in no go pitch diameter.
  • Single end configuration for specific applications.
  • TiN coating/finish for enhanced wear resistance.