Learn More
MSC GF Gage Plug Thread Gage, #4-48 Thread, 3B Class, Double End Go/No Go
Catalog No. 5000535010
Encompass
This GF Gage Plug Thread Gage is designed for precise measurement of #4-48 threads with a 3B class fit. It features a double-ended configuration, providing both 'Go' and 'No Go' sides for comprehensive thread inspection. Constructed from hardened tool steel, this gage ensures durability and accuracy for laboratory and industrial applications.
  • Go pitch diameter: 0.0985 in
  • No Go pitch diameter: 0.1008 in
  • Handle included
  • Constructed from hardened tool steel
  • 3B class for thread fit
  • Designed for #4-48 thread size
  • Double-ended configuration
  • Manufactured in the United States
Catalog No. 50-005-35010 Supplier MSC Supplier No. 55310890
This GF Gage Plug Thread Gage is designed for precise measurement of #4-48 threads with a 3B class fit. It features a double-ended configuration, providing both 'Go' and 'No Go' sides for comprehensive thread inspection. Constructed from hardened tool steel, this gage ensures durability and accuracy for laboratory and industrial applications.
  • Go pitch diameter: 0.0985 in
  • No Go pitch diameter: 0.1008 in
  • Handle included
  • Constructed from hardened tool steel
  • 3B class for thread fit
  • Designed for #4-48 thread size
  • Double-ended configuration
  • Manufactured in the United States