Learn More
MSC GF Gage Plug Thread Gage, #6-32 Thread, 2B/3B Class, Single End Go
Catalog No. 5000512147
Encompass
This plug thread gage from GF Gage is designed to accurately verify #6-32 threads. It features a single end with a 'Go' designation, ensuring compliance with both 2B and 3B thread classes. Manufactured from hardened tool steel, this gage provides reliable performance for precision thread measurement.
  • Verifies #6-32 threads for accuracy.
  • Suitable for 2B and 3B thread classifications.
  • Single end 'Go' design checks minimum thread size.
  • Constructed from hardened tool steel for extended durability.
  • Supplied with a certificate of compliance.
Catalog No. 50-005-12147 Supplier MSC Supplier No. 55306310
This plug thread gage from GF Gage is designed to accurately verify #6-32 threads. It features a single end with a 'Go' designation, ensuring compliance with both 2B and 3B thread classes. Manufactured from hardened tool steel, this gage provides reliable performance for precision thread measurement.
  • Verifies #6-32 threads for accuracy.
  • Suitable for 2B and 3B thread classifications.
  • Single end 'Go' design checks minimum thread size.
  • Constructed from hardened tool steel for extended durability.
  • Supplied with a certificate of compliance.