Learn More
MSC GF Gage Plug Thread Gage, 7/16-20, 2B/3B Class, Single End, Go
Catalog No. 5000514782
Encompass
This plug thread gage is engineered for precise inspection of internal threads, specifically for 7/16-20 thread sizes. It ensures quality control by verifying "Go" conditions for both 2B and 3B classifications.
  • Designed for precise inspection of 7/16-20 threads.
  • Verifies "Go" conditions for 2B and 3B thread classifications.
  • Constructed from hardened tool steel for extended durability.
  • Single end design.
  • Features a Go pitch diameter of 0.4050 "
Catalog No. 50-005-14782 Supplier MSC Supplier No. 55306674
This plug thread gage is engineered for precise inspection of internal threads, specifically for 7/16-20 thread sizes. It ensures quality control by verifying "Go" conditions for both 2B and 3B classifications.
  • Designed for precise inspection of 7/16-20 threads.
  • Verifies "Go" conditions for 2B and 3B thread classifications.
  • Constructed from hardened tool steel for extended durability.
  • Single end design.
  • Features a Go pitch diameter of 0.4050 "