Learn More
MSC GF Gage Plug Thread Gage, 7/16-28 Thread, 2B Class, Double End, Go & No Go
Catalog No. 5000536850
Encompass
This plug thread gage is designed for inspecting 7/16-28 threads. It features a double-ended design with both "Go" and "No Go" functions, providing comprehensive thread inspection. Made from hardened tool steel, it ensures durability and precision for consistent performance.
  • Inspects 7/16-28 threads
  • Double-ended with "Go" and "No Go" functions
  • 2B class for accurate measurement
  • Includes a handle for ease of use
  • Go pitch diameter is 0.4143 inches
  • No go pitch diameter is 0.41890 inches
  • Made from hardened tool steel for durability
  • UNEF compliant
Catalog No. 50-005-36850 Supplier MSC Supplier No. 55310171
This plug thread gage is designed for inspecting 7/16-28 threads. It features a double-ended design with both "Go" and "No Go" functions, providing comprehensive thread inspection. Made from hardened tool steel, it ensures durability and precision for consistent performance.
  • Inspects 7/16-28 threads
  • Double-ended with "Go" and "No Go" functions
  • 2B class for accurate measurement
  • Includes a handle for ease of use
  • Go pitch diameter is 0.4143 inches
  • No go pitch diameter is 0.41890 inches
  • Made from hardened tool steel for durability
  • UNEF compliant