Learn More
MSC GF Gage Plug Thread Gage, M8x1.25 Thread, 6H Class, Single End, No Go
Catalog No. 5000511494
Encompass
This plug thread gage is designed for M8x1.25 threads with a 6H class. It functions as a single-end, 'No Go' gage, providing precise measurements for quality control. Constructed from hardened tool steel, it ensures durability and accuracy in various industrial applications.
  • No go functionality
  • Single end design
  • M8x1.25 thread size
  • Constructed from hardened tool steel
  • 6H classification
  • 7.3500 mm No Go pitch diameter
Catalog No. 50-005-11494 Supplier MSC Supplier No. 55318893
This plug thread gage is designed for M8x1.25 threads with a 6H class. It functions as a single-end, 'No Go' gage, providing precise measurements for quality control. Constructed from hardened tool steel, it ensures durability and accuracy in various industrial applications.
  • No go functionality
  • Single end design
  • M8x1.25 thread size
  • Constructed from hardened tool steel
  • 6H classification
  • 7.3500 mm No Go pitch diameter