Learn More
MSC GF Gage Plug Thread Insert Gage, 7/16-20 Thread, STI, Class 2B/3B, Go
Catalog No. 5000512831
Encompass
This plug thread insert go gage is designed for precision measurement of 7/16-20 screw thread inserts. Constructed from durable steel, it ensures accuracy and reliability for quality control applications.
  • Single end without handle.
  • Designed for screw thread inserts (STI).
  • Measures pitch diameter for both 2B and 3B class of fit at .4700.
Catalog No. 50-005-12831 Supplier MSC Supplier No. 55315055
This plug thread insert go gage is designed for precision measurement of 7/16-20 screw thread inserts. Constructed from durable steel, it ensures accuracy and reliability for quality control applications.
  • Single end without handle.
  • Designed for screw thread inserts (STI).
  • Measures pitch diameter for both 2B and 3B class of fit at .4700.