Learn More
MSC GF Gage Plug Thread Insert Go Gage, 1/4-20 Thread, STI 2B/3B, Steel
Catalog No. 5000528615
Encompass
This plug thread insert go gage is designed for precise measurement of 1/4-20 threads. It is manufactured from durable steel and features Screw Thread Insert (STI) classification 2B/3B, ensuring accurate gauging for specific thread requirements. The gage is single-ended and supplied without a handle.
  • Designed for 1/4-20 thread size
  • Features a go/no go classification
  • Suitable for screw thread insert (STI) applications
  • Complies with 2B/3B classification standards
  • Constructed from steel for durability
  • Single-ended design
  • Handle is not included
  • Supplied with a certificate of conformance
Catalog No. 50-005-28615 Supplier MSC Supplier No. 55314934
This plug thread insert go gage is designed for precise measurement of 1/4-20 threads. It is manufactured from durable steel and features Screw Thread Insert (STI) classification 2B/3B, ensuring accurate gauging for specific thread requirements. The gage is single-ended and supplied without a handle.
  • Designed for 1/4-20 thread size
  • Features a go/no go classification
  • Suitable for screw thread insert (STI) applications
  • Complies with 2B/3B classification standards
  • Constructed from steel for durability
  • Single-ended design
  • Handle is not included
  • Supplied with a certificate of conformance