Learn More
MSC GF Gage Plug Thread Insert Go/No Go Gage, #2-56 Thread, Class 3B, Steel, Double End
Catalog No. 5000531238
Encompass
This plug thread insert go/no go gage is designed for #2-56 thread applications. It is constructed from steel and features a Class 3B classification for Screw Thread Insert (STI) measurements. The gage features a double-ended design and includes a handle for ease of use.
  • Double end design with an included handle.
  • Designed for #2-56 thread applications.
  • Constructed from steel for durability.
  • Features a Class 3B classification for Screw Thread Insert (STI) measurements.
  • Provides go/no go functionality for precise thread inspection.
Catalog No. 50-005-31238 Supplier MSC Supplier No. 55316012
This plug thread insert go/no go gage is designed for #2-56 thread applications. It is constructed from steel and features a Class 3B classification for Screw Thread Insert (STI) measurements. The gage features a double-ended design and includes a handle for ease of use.
  • Double end design with an included handle.
  • Designed for #2-56 thread applications.
  • Constructed from steel for durability.
  • Features a Class 3B classification for Screw Thread Insert (STI) measurements.
  • Provides go/no go functionality for precise thread inspection.