Learn More
MSC GF Gage Plug Thread Insert Go/No Go Gage, 3/8-16, Steel, STI Class 3B
Catalog No. 5000527818
Encompass
This GF Gage is a double end plug thread insert go/no go gage. Crafted from durable steel, it is designed for precise 3/8-16 thread applications with a Class 3B classification and includes a handle for ease of use.
  • Go/no go functionality
  • Designed for 3/8-16 thread applications
  • Handle size 2
  • Class 3B classification
  • Screw thread insert (STI) type
  • Includes a handle
  • Constructed from steel
  • Double-ended design
Catalog No. 50-005-27818 Supplier MSC Supplier No. 55316210
This GF Gage is a double end plug thread insert go/no go gage. Crafted from durable steel, it is designed for precise 3/8-16 thread applications with a Class 3B classification and includes a handle for ease of use.
  • Go/no go functionality
  • Designed for 3/8-16 thread applications
  • Handle size 2
  • Class 3B classification
  • Screw thread insert (STI) type
  • Includes a handle
  • Constructed from steel
  • Double-ended design