Learn More
MSC GF Gage Plug Thread Insert Go/No Go Gage, 3/8-24 Thread, Steel, Class 3B, Double End
Catalog No. 5000531612
Encompass
This GF Gage is a Plug Thread Insert Go/No Go Gage designed for precise inspection of 3/8-24 threads. Constructed from durable steel, it adheres to Class 3B standards for accuracy. The double-ended design, including a handle, ensures efficient and reliable thread verification for Screw Thread Insert (STI) applications.
  • Go and No Go functionality for accurate thread inspection.
  • Designed for 3/8-24 thread size.
  • Conforms to Class 3B classification.
  • Specifically for Screw Thread Insert (STI) applications.
  • Constructed from durable steel for longevity.
  • Double-ended design with included handle for ease of use.
  • Supplied with a Certificate of Compliance.
Catalog No. 50-005-31612 Supplier MSC Supplier No. 55316236
This GF Gage is a Plug Thread Insert Go/No Go Gage designed for precise inspection of 3/8-24 threads. Constructed from durable steel, it adheres to Class 3B standards for accuracy. The double-ended design, including a handle, ensures efficient and reliable thread verification for Screw Thread Insert (STI) applications.
  • Go and No Go functionality for accurate thread inspection.
  • Designed for 3/8-24 thread size.
  • Conforms to Class 3B classification.
  • Specifically for Screw Thread Insert (STI) applications.
  • Constructed from durable steel for longevity.
  • Double-ended design with included handle for ease of use.
  • Supplied with a Certificate of Compliance.