Learn More
MSC GF Gage Plug Thread Insert Go/No Go Gage, #8-32 Thread, Class 3B, Steel
Catalog No. 5000528330
Encompass
This GF Gage Plug Thread Insert Go/No Go Gage is designed for accurately checking #8-32 screw threads with a Class 3B classification. Constructed from durable steel and featuring a double-ended design, this gage ensures precise measurement and comes with a certificate of compliance.
  • Go/no go functionality
  • #8-32 thread size
  • Class 3B thread classification
  • Designed for screw thread inserts (STI)
  • Includes a handle
  • Made from steel
  • Double-ended design
  • Comes with a certificate of compliance
Catalog No. 50-005-28330 Supplier MSC Supplier No. 55316079
This GF Gage Plug Thread Insert Go/No Go Gage is designed for accurately checking #8-32 screw threads with a Class 3B classification. Constructed from durable steel and featuring a double-ended design, this gage ensures precise measurement and comes with a certificate of compliance.
  • Go/no go functionality
  • #8-32 thread size
  • Class 3B thread classification
  • Designed for screw thread inserts (STI)
  • Includes a handle
  • Made from steel
  • Double-ended design
  • Comes with a certificate of compliance