Learn More
MSC GF Gage Ring Gage, #2-56 Thread, UNC, Class 3A, Go
Catalog No. 5000531096
Encompass
The GF Gage Ring Gage is a precision tool designed for inspecting threaded parts. It ensures accuracy by checking the external thread dimensions against specific standards.
  • Designed to check "Go" specifications for external threads.
  • Features a #2-56 thread size.
  • Utilizes a UNC thread type.
  • Meets Class 3A classification standards.
  • Provided as a single ring for focused inspection.
Catalog No. 50-005-31096 Supplier MSC Supplier No. 55321491
The GF Gage Ring Gage is a precision tool designed for inspecting threaded parts. It ensures accuracy by checking the external thread dimensions against specific standards.
  • Designed to check "Go" specifications for external threads.
  • Features a #2-56 thread size.
  • Utilizes a UNC thread type.
  • Meets Class 3A classification standards.
  • Provided as a single ring for focused inspection.