Learn More
MSC GF Gage Ring Gage Holder, 9/16-3/4 in Dia Range, 4 Holder Size
Catalog No. 5000527726
Encompass
This ring gage holder accommodates ring diameters from 9/16 to 3/4 in (14.29-19.05 mm) and features a 4 holder size. It is manufactured in the United States.
  • Accommodates ring diameters from 9/16 to 3/4 in.
  • Features a 4 holder size.
  • Manufactured in the United States.
  • Designed for secure holding of ring gages during inspection or calibration.
Catalog No. 50-005-27726 Supplier MSC Supplier No. 55338891
This ring gage holder accommodates ring diameters from 9/16 to 3/4 in (14.29-19.05 mm) and features a 4 holder size. It is manufactured in the United States.
  • Accommodates ring diameters from 9/16 to 3/4 in.
  • Features a 4 holder size.
  • Manufactured in the United States.
  • Designed for secure holding of ring gages during inspection or calibration.