Learn More
MSC GF Gage STI Go/No Go Gage, 1/4-28, Class 2B, Steel
Catalog No. 5000531095
Encompass
This plug thread insert Go/No Go gage is designed for 1/4-28 threads. Constructed from steel, it features an STI Class 2B classification and a double-ended design, including a handle for ease of use.
  • Go/No Go functionality
  • 1/4-28 thread size
  • Class 2B classification
  • Screw thread insert (STI) type
  • Includes a handle
  • Constructed from steel
  • Double-ended design
  • Comes with a general certificate of conformance
  • NIST serialized certificate of calibration available upon request for an additional fee
  • One unit ordered provides a 3-piece set
Catalog No. 50-005-31095 Supplier MSC Supplier No. 55315857
This plug thread insert Go/No Go gage is designed for 1/4-28 threads. Constructed from steel, it features an STI Class 2B classification and a double-ended design, including a handle for ease of use.
  • Go/No Go functionality
  • 1/4-28 thread size
  • Class 2B classification
  • Screw thread insert (STI) type
  • Includes a handle
  • Constructed from steel
  • Double-ended design
  • Comes with a general certificate of conformance
  • NIST serialized certificate of calibration available upon request for an additional fee
  • One unit ordered provides a 3-piece set