Learn More
MSC GF Gage Threaded Pipe Ring, 1/2-14, NPTF, Class L2
Catalog No. 5000531276
Encompass
This threaded pipe ring is designed for gauging threads. It is classified as L2, indicating its specific tolerance or application.
  • Designed for NPTF thread type.
  • Features a 1/2-14 inch thread size.
  • Class L2 classification for specific gauging requirements.
Catalog No. 50-005-31276 Supplier MSC Supplier No. 55337299
This threaded pipe ring is designed for gauging threads. It is classified as L2, indicating its specific tolerance or application.
  • Designed for NPTF thread type.
  • Features a 1/2-14 inch thread size.
  • Class L2 classification for specific gauging requirements.