Learn More
MSC GF Gage Threaded Ring Gage, 1-1/8-12 Thread, Class 3A, No Go
Catalog No. 5000527634
Encompass
This threaded ring gage is designed for precise measurement of threads. It features a 1-1/8-12 thread size and a Class 3A classification, functioning as a 'No Go' gage to ensure proper thread tolerances.
  • No Go functionality
  • 1-1/8-12 thread size
  • Class 3A classification
  • Single ring design
Catalog No. 50-005-27634 Supplier MSC Supplier No. 55324776
This threaded ring gage is designed for precise measurement of threads. It features a 1-1/8-12 thread size and a Class 3A classification, functioning as a 'No Go' gage to ensure proper thread tolerances.
  • No Go functionality
  • 1-1/8-12 thread size
  • Class 3A classification
  • Single ring design