Learn More
MSC GF Gage Threaded Ring Gage, 1/2-28 Thread, UNEF, Class 3A, Go
Catalog No. 5000535148
Encompass
This threaded ring gage is designed for inspecting 1/2-28 UNEF threads with a Class 3A classification. It functions as a "Go" gage, specifically used to verify the upper limit of thread tolerance.
  • Go gage
  • 1/2-28 thread size
  • UNEF thread type
  • Class 3A classification
  • Single ring design
  • Checks the upper limit of thread tolerance
Catalog No. 50-005-35148 Supplier MSC Supplier No. 55322093
This threaded ring gage is designed for inspecting 1/2-28 UNEF threads with a Class 3A classification. It functions as a "Go" gage, specifically used to verify the upper limit of thread tolerance.
  • Go gage
  • 1/2-28 thread size
  • UNEF thread type
  • Class 3A classification
  • Single ring design
  • Checks the upper limit of thread tolerance