Learn More
MSC GF Gage Threaded Ring Gage, 1/4-28 Thread, UNF, Class 3A, Go
Catalog No. 5000535216
Encompass
This product is a "Go" threaded ring gage designed for inspecting 1/4-28 UNF threads with a Class 3A fit.
  • Designed as a go gage
  • 1/4-28 thread size
  • UNF thread type
  • Single ring design
  • Class 3A classification
  • Made in the United States
Catalog No. 50-005-35216 Supplier MSC Supplier No. 55321830
This product is a "Go" threaded ring gage designed for inspecting 1/4-28 UNF threads with a Class 3A fit.
  • Designed as a go gage
  • 1/4-28 thread size
  • UNF thread type
  • Single ring design
  • Class 3A classification
  • Made in the United States