Learn More
MSC GF Gage Threaded Ring Gage, #10-24 Thread, UNC, Class 2A, Go/No Go
Catalog No. 5000535171
Encompass
This threaded ring gage is designed for inspecting #10-24 UNC threads with a Class 2A classification. It features both 'Go' and 'No Go' functionalities to ensure precise quality control for manufacturing processes, verifying that parts meet specified thread tolerances.
  • Allows for inspection of #10-24 UNC threads with a 2A classification.
  • Features "Go" and "No Go" functionalities.
  • Crucial for quality control in manufacturing processes.
  • Ensures parts meet specified thread tolerances.
  • Double ring design.
Catalog No. 50-005-35171 Supplier MSC Supplier No. 55325211
This threaded ring gage is designed for inspecting #10-24 UNC threads with a Class 2A classification. It features both 'Go' and 'No Go' functionalities to ensure precise quality control for manufacturing processes, verifying that parts meet specified thread tolerances.
  • Allows for inspection of #10-24 UNC threads with a 2A classification.
  • Features "Go" and "No Go" functionalities.
  • Crucial for quality control in manufacturing processes.
  • Ensures parts meet specified thread tolerances.
  • Double ring design.