Learn More
MSC GF Gage Threaded Ring Gage, #10-32 UNF, 2A Go, Tool Steel
Catalog No. 5000527817
Encompass
This threaded ring gage is designed for the precise inspection of #10-32 UNF threads. It functions as a 'Go' gage, ensuring that components meet the specified 2A classification for quality control in manufacturing and assembly processes.
  • Ensures accurate measurement of #10-32 UNF threads.
  • Conforms to 2A classification standards for quality assurance.
  • Manufactured from durable tool steel for extended service life.
  • Single ring design for specific gaging requirements.
Catalog No. 50-005-27817 Supplier MSC Supplier No. 55320592
This threaded ring gage is designed for the precise inspection of #10-32 UNF threads. It functions as a 'Go' gage, ensuring that components meet the specified 2A classification for quality control in manufacturing and assembly processes.
  • Ensures accurate measurement of #10-32 UNF threads.
  • Conforms to 2A classification standards for quality assurance.
  • Manufactured from durable tool steel for extended service life.
  • Single ring design for specific gaging requirements.