Learn More
MSC GF Gage Threaded Ring Gage, #2-56 Thread, UNC, Class 3A, Go/No Go
Catalog No. 5000537479
Encompass
This Threaded Ring Gage is designed for #2-56 UNC threads, featuring a Class 3A tolerance and including both Go and No Go measuring ends to ensure precise inspection of threaded components.
  • Designed for #2-56 UNC threads.
  • Features class 3A tolerance.
  • Includes both go and no go measuring ends.
  • Ensures precise inspection of threaded components.
  • Double ring type for comprehensive gaging.
  • Manufactured in the United States.
Catalog No. 50-005-37479 Supplier MSC Supplier No. 55326136
This Threaded Ring Gage is designed for #2-56 UNC threads, featuring a Class 3A tolerance and including both Go and No Go measuring ends to ensure precise inspection of threaded components.
  • Designed for #2-56 UNC threads.
  • Features class 3A tolerance.
  • Includes both go and no go measuring ends.
  • Ensures precise inspection of threaded components.
  • Double ring type for comprehensive gaging.
  • Manufactured in the United States.